2-Chloro-3-nitropyridin-4-amine

Supplier: Apollo Scientific
Warning

Synonyms: 4-Amino-2-chloro-3-nitropyridine

OR15937-500MG
APOSOR15937-500MGEA 21.9 EUR
APOSOR15937-500MG
2-Chloro-3-nitropyridin-4-amine
2-Chloro-3-nitropyridin-4-amine
Formula: C₅H₄ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07368898
CAS Number: 2789-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR