4-(Prop-1-ylamino)piperidine, N1-BOC protected 97%

Supplier: Apollo Scientific

Synonyms: tert-Butyl 4-(propylamino)piperidine-1-carboxylate

OR16031-5G
APOSOR16031-5GEA 87 EUR
APOSOR16031-5G
4-(Prop-1-ylamino)piperidine, N1-BOC protected 97%
4-(Prop-1-ylamino)piperidine, N1-BOC protected
Formula: C₁₃H₂₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07367771
CAS Number: 301225-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR