3-Hydroxy-2-nitro-6-picoline 95%

Supplier: Apollo Scientific
Warning

OR16078-100G OR16078-5G OR16078-25G
APOSOR16078-100GEA 644 EUR
APOSOR16078-100G APOSOR16078-5G APOSOR16078-25G
3-Hydroxy-2-nitro-6-picoline 95%
3-Hydroxy-2-nitro-6-picoline
Formula: C₆H₆N₂O₃
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00006259
CAS Number: 15128-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR