3-Chloro-2-methylpropene

Supplier: Apollo Scientific
Danger

OR16423-250G OR16423-1KG
APOSOR16423-250GEA 0 EUR
APOSOR16423-250G APOSOR16423-1KG
3-Chloro-2-methylpropene
3-Chloro-2-methylpropene
Formula: C₄H₇Cl
Boiling Pt: 71…72 °C
Melting Pt: –80 °C
Density: 0,925 g/cm³ (25 °C)
Flash Pt: –10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000953
CAS Number: 563-47-3
EINECS: 209-251-2
UN: 2554
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR