Tetrabutylammonium nitrate

Supplier: Apollo Scientific
Danger

OR16429-100G OR16429-500G
APOSOR16429-100GEA 209 EUR
APOSOR16429-100G APOSOR16429-500G
Tetrabutylammonium nitrate
Tetrabutylammonium nitrate
Formula: C₁₆H₃₆N₂O₃
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00043202
CAS Number: 1941-27-1
EINECS: 217-726-0
UN: 1479
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR