Glycolaldehyde diethyl acetal

Supplier: Apollo Scientific
Warning

OR16597-100G OR16597-25G OR16597-5G
APOSOR16597-100GEA 299 EUR
APOSOR16597-100G APOSOR16597-25G APOSOR16597-5G
Glycolaldehyde diethyl acetal
Glycolaldehyde diethyl acetal
Formula: HOCH₂CH(OCH₂CH₃)₂
Boiling Pt: 166…167 °C
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051486
CAS Number: 621-63-6
EINECS: 210-697-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR