3-Bromo-2-methylbenzaldehyde 97%

Supplier: Apollo Scientific
Warning

OR16673-1G OR16673-250MG OR16673-5G
APOSOR16673-1GEA 24.8 EUR
APOSOR16673-1G APOSOR16673-250MG APOSOR16673-5G
3-Bromo-2-methylbenzaldehyde 97%
3-Bromo-2-methylbenzaldehyde
Formula: C₈H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD11847533
CAS Number: 83647-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR