6-Benzyloxyindole-3-carbaldehyde 98%

Supplier: Apollo Scientific
Warning

OR1676-1G
APOSOR1676-1GEA 29 EUR
APOSOR1676-1G
6-Benzyloxyindole-3-carbaldehyde 98%
6-Benzyloxyindole-3-carbaldehyde
Formula: C₁₆H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00056931
CAS Number: 92855-64-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR