tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate 98%

Supplier: Apollo Scientific
Warning

OR1692-1G OR1692-5G
APOSOR1692-1GEA 30.6 EUR
APOSOR1692-1G APOSOR1692-5G
tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate 98%
tert-Butyl-3-(hydroxymethyl)-5-nitro-1H-indole-1-carboxylate
Formula: C₁₄H₁₆N₂O₅
Storage Temperature: Ambient
MDL Number: MFCD05864712
CAS Number: 914349-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR