2-Hydroxyethylurea

Supplier: Apollo Scientific

Synonyms: 1-(2-Hydroxyethyl)urea

OR17003-100G OR17003-250G OR17003-500G
APOSOR17003-100GEA 132 EUR
APOSOR17003-100G APOSOR17003-250G APOSOR17003-500G
2-Hydroxyethylurea
2-Hydroxyethylurea
Formula: C₃H₈N₂O₂
Melting Pt: 164…169 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00059080
CAS Number: 2078-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR