7-Methoxy-2,3-dihydro-1H-inden-1-one

Supplier: Apollo Scientific
Warning

OR17401-1G OR17401-5G OR17401-25G
APOSOR17401-1GEA 33.5 EUR
APOSOR17401-1G APOSOR17401-5G APOSOR17401-25G
7-Methoxy-2,3-dihydro-1H-inden-1-one
7-Methoxy-2,3-dihydro-1H-inden-1-one
Formula: C₁₀H₁₀O₂
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD07784339
CAS Number: 34985-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR