AMP (2-Amino-2-methylpropanol) 95%

Supplier: Apollo Scientific
Warning

OR1740T-1KG OR1740T-500G
APOSOR1740T-1KGEA 106 EUR
APOSOR1740T-1KG APOSOR1740T-500G
AMP (2-Amino-2-methylpropanol) 95%
AMP (2-Amino-2-methylpropanol)

Used for the preparation of buffer solutions, suitable for the determination of alkaline phosphatase.

Formula: (CH₃)₂C(NH₂)CH₂OH
Boiling Pt: 164…167 °C
Melting Pt: 24…32 °C
Density: 0,934 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00008051
CAS Number: 124-68-5
EINECS: 204-709-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR