2-Amino-6-bromobenzonitrile 97%

Supplier: Apollo Scientific
Warning

OR17502-5G OR17502-100G OR17502-25G
APOSOR17502-5GEA 24.8 EUR
APOSOR17502-5G APOSOR17502-100G APOSOR17502-25G
2-Amino-6-bromobenzonitrile 97%
2-Amino-6-bromobenzonitrile
Formula: C₇H₅BrN₂
Melting Pt: 152…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834769
CAS Number: 77326-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR