4-Bromo-2,6-dinitrotoluene

Supplier: Apollo Scientific
Warning

OR17614-1G OR17614-5G
APOSOR17614-1GEA 36.4 EUR
APOSOR17614-1G APOSOR17614-5G
4-Bromo-2,6-dinitrotoluene
4-Bromo-2,6-dinitrotoluene
Formula: C₇H₅BrN₂O₄
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD07357280
CAS Number: 95192-64-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR