1-(4-Chlorophenyl)ethanamine, Technical Grade

Supplier: Apollo Scientific
Danger

OR17821-1G OR17821-5G OR17821-10G
APOSOR17821-1GEA 21.9 EUR
APOSOR17821-1G APOSOR17821-5G APOSOR17821-10G
1-(4-Chlorophenyl)ethanamine, Technical Grade
1-(4-Chlorophenyl)ethanamine
Formula: C₈H₁₀ClN
Boiling Pt: 82,4 °C/2.8mmHg
Density: 1,08 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044766
CAS Number: 6299-02-1
EINECS: 228-576-0
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR