4-Bromo-2,6-dimethylpyridine

Supplier: Apollo Scientific
Warning

OR17852-1G
APOSOR17852-1GEA 21.9 EUR
APOSOR17852-1G
4-Bromo-2,6-dimethylpyridine
4-Bromo-2,6-dimethylpyridine
Formula: C₇H₈BrN
Boiling Pt: 81 °C/14mmHg
Melting Pt: 32…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00671475
CAS Number: 5093-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR