5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan

Supplier: Apollo Scientific
Warning

OR17915-5G
APOSOR17915-5GEA 190 EUR
APOSOR17915-5G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Formula: C₁₄H₁₇BO₃
Boiling Pt: 90…120 °C/0mmHg
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD04115374
CAS Number: 519054-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR