2-Amino-3-methylphenylacetic acid

Supplier: Apollo Scientific
Warning

OR18018-1G OR18018-250MG
APOSOR18018-1GEA 2970 EUR
APOSOR18018-1G APOSOR18018-250MG
2-Amino-3-methylphenylacetic acid
2-Amino-3-methylphenylacetic acid
Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD16606085
CAS Number: 1334405-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR