5-Chloro-2-ethoxy-nicotinic acid

Supplier: Apollo Scientific
Warning

OR18046-1G OR18046-250MG OR18046-5G
APOSOR18046-1GEA 31.9 EUR
APOSOR18046-1G APOSOR18046-250MG APOSOR18046-5G
5-Chloro-2-ethoxy-nicotinic acid
5-Chloro-2-ethoxy-nicotinic acid
Formula: C₈H₈ClNO₃
Melting Pt: 115,5…117.5 °C
Storage Temperature: Ambient
MDL Number: MFCD07403147
CAS Number: 68359-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR