2-(2-Hydroxyphenyl)benzothiazole 98%

Supplier: Apollo Scientific
Warning

OR18303-25G
APOSOR18303-25GEA 87 EUR
APOSOR18303-25G
2-(2-Hydroxyphenyl)benzothiazole 98%
2-(2-Hydroxyphenyl)benzothiazole
Formula: C₁₃H₉NOS
Boiling Pt: 190…193 °C/3mmHg
Melting Pt: 128…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00022869
CAS Number: 3411-95-8
EINECS: 222-299-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR