1-Methyl-4-[4-(1H-pyrazol-3-yl)phenyl]sulphonyl-piperazine

Supplier: Apollo Scientific

OR183441-1G
APOSOR183441-1GEA 219 EUR
APOSOR183441-1G
1-Methyl-4-[4-(1H-pyrazol-3-yl)phenyl]sulphonyl-piperazine
1-Methyl-4-[4-(1H-pyrazol-3-yl)phenyl]sulphonyl-piperazine
Formula: C₁₄H₁₈N₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD07774266
CAS Number: 1019012-25-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR