4-Chloro-3,5-dinitrobenzoic acid

Supplier: Apollo Scientific
Warning

Synonyms: 3,5-Dinitro-4-chlorobenzoic acid

OR18359-500G
APOSOR18359-500GEA 98.6 EUR
APOSOR18359-500G
4-Chloro-3,5-dinitrobenzoic acid
4-Chloro-3,5-dinitrobenzoic acid
Formula: C₇H₃ClN₂O₆
MW: 246.56 g/mol
Boiling Pt: 315 °C
Melting Pt: 159…162 °C
Flash Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00007080
CAS Number: 118-97-8
EINECS: 204-209-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR