4-(Benzyloxy)cyclohexanone 96%

Supplier: Apollo Scientific
Warning

OR18843-10G OR18843-5G OR18843-25G
APOSOR18843-10GEA 137 EUR
APOSOR18843-10G APOSOR18843-5G APOSOR18843-25G
4-(Benzyloxy)cyclohexanone 96%
4-(Benzyloxy)cyclohexanone
Formula: C₁₃H₁₆O₂
Boiling Pt: 102…103 °C/0,15mmHg
Storage Temperature: Ambient
MDL Number: MFCD10697867
CAS Number: 2987-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR