1-(tert-Butoxycarbonyl)-3-phenylpiperidine-3-carboxylic acid

Supplier: Apollo Scientific
Danger

OR200004-100MG OR200004-250MG
APOSOR200004-100MGEA 661 EUR
APOSOR200004-100MG APOSOR200004-250MG
1-(tert-Butoxycarbonyl)-3-phenylpiperidine-3-carboxylic acid
1-(tert-Butoxycarbonyl)-3-phenylpiperidine-3-carboxylic acid
Formula: C₁₇H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD11223632
CAS Number: 1203685-64-6
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR