1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid

Supplier: Apollo Scientific

Synonyms: 1-[4-(Methylsulphonyl)phenyl]-5-oxo-3-pyrrolidinecarboxylic acid

OR200035-1G OR200035-5G OR200035-10G
APOSOR200035-1GEA 116 EUR
APOSOR200035-1G APOSOR200035-5G APOSOR200035-10G
1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid
1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid
Formula: C₁₂H₁₃NO₃S
MW: 251.31 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD08444946
CAS Number: 63674-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR