3-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid 98%

Supplier: Apollo Scientific
Warning

OR2098-250MG
APOSOR2098-250MGEA 21.9 EUR
APOSOR2098-250MG
3-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid 98%
3-(2,5-Dimethylphenylcarbamoyl)phenylboronic acid
Formula: C₁₅H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD09027263
CAS Number: 957061-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR