5-Bromo-2-methoxy-4-pyridineboronic acid 98%

Supplier: Apollo Scientific

OR2099-1G
APOSOR2099-1GEA 78.3 EUR
APOSOR2099-1G
5-Bromo-2-methoxy-4-pyridineboronic acid 98%
5-Bromo-2-methoxy-4-pyridineboronic acid
Formula: C₆H₇BBrNO₃
Storage Temperature: Ambient
MDL Number: MFCD07781223
CAS Number: 957060-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR