5-Methoxypyridine-3-boronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid

OR2109-1G OR2109-5G
APOSOR2109-1GEA 26.1 EUR
APOSOR2109-1G APOSOR2109-5G
5-Methoxypyridine-3-boronic acid
5-Methoxypyridine-3-boronic acid
Formula: C₆H₈BNO₃
MW: 152.95 g/mol
Melting Pt: 224…227 °C
Storage Temperature: Ambient
MDL Number: MFCD03425977
CAS Number: 850991-69-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR