1-Bromo-4-butoxybenzene 97%

Supplier: Apollo Scientific
Warning

OR21904-1G OR21904-5G
APOSOR21904-1GEA 21.9 EUR
APOSOR21904-1G APOSOR21904-5G
1-Bromo-4-butoxybenzene 97%
1-Bromo-4-butoxybenzene
Formula: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD00037100
CAS Number: 39969-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR