3,5-Di-tert-butyl-4-hydroxybenzonitrile 97%

Supplier: Apollo Scientific
Warning

OR21941-1G OR21941-5G
APOSOR21941-1GEA 31.9 EUR
APOSOR21941-1G APOSOR21941-5G
3,5-Di-tert-butyl-4-hydroxybenzonitrile 97%
3,5-Di-tert-butyl-4-hydroxybenzonitrile
Formula: C₁₅H₂₁NO
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00156137
CAS Number: 1988-88-1
UN: 3439
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR