4-Oxopent-2-enoic acid

Supplier: Apollo Scientific
Warning

OR22045-1G OR22045-50MG OR22045-250MG OR22045-500MG OR22045-2.5G
APOSOR22045-1GEA 340 EUR
APOSOR22045-1G APOSOR22045-50MG APOSOR22045-250MG APOSOR22045-500MG APOSOR22045-2.5G
4-Oxopent-2-enoic acid
4-Oxopent-2-enoic acid
Formula: C₅H₆O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00014016
CAS Number: 4743-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR