(S)-2-Aminopropanamide hydrochloride

Supplier: Apollo Scientific
Warning

Synonyms: H-Ala-NH2.HCl, (S)-2-Aminopropionamide Hydrochloride, L-Alanamine Hydrochloride, H-Ala-NH₂·HCl, L-Alaninamide hydrochloride

OR22615-100G OR22615-25G
APOSOR22615-100GEA 94.4 EUR
APOSOR22615-100G APOSOR22615-25G
(S)-2-Aminopropanamide hydrochloride
(S)-2-Aminopropanamide hydrochloride
Formula: C₃H₉ClN₂O
MW: 124.57 g/mol
Melting Pt: 212…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00066145
CAS Number: 33208-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR