3-Methylbiphenyl

Supplier: Apollo Scientific
Warning

OR22658-1G
APOSOR22658-1GEA 21.9 EUR
APOSOR22658-1G
3-Methylbiphenyl
3-Methylbiphenyl
Formula: C₁₃H₁₂
Boiling Pt: 272 °C
Melting Pt: 44320 °C
Storage Temperature: Ambient
MDL Number: MFCD00008533
CAS Number: 643-93-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR