tert-Butyl thiophen-2-ylcarbamate

Supplier: Apollo Scientific
Warning

OR22958-1G OR22958-5G OR22958-10G
APOSOR22958-1GEA 21.9 EUR
APOSOR22958-1G APOSOR22958-5G APOSOR22958-10G
tert-Butyl thiophen-2-ylcarbamate
tert-Butyl thiophen-2-ylcarbamate
Formula: C₉H₁₃NO₂S
Melting Pt: 151…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02677742
CAS Number: 56267-50-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR