5-Bromo-2,4-dimethyl-1,3-thiazole

Supplier: Apollo Scientific
Warning

OR23084-1G
APOSOR23084-1GEA 49.3 EUR
APOSOR23084-1G
5-Bromo-2,4-dimethyl-1,3-thiazole
5-Bromo-2,4-dimethyl-1,3-thiazole
Formula: C₅H₆BrNS
Boiling Pt: 114…120 °C/15mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD03086099
CAS Number: 28599-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR