4-(4-Methylpiperazin-1-yl)benzaldehyde

Supplier: Apollo Scientific
Warning

OR23309-25G
APOSOR23309-25GEA 127 EUR
APOSOR23309-25G
4-(4-Methylpiperazin-1-yl)benzaldehyde
4-(4-Methylpiperazin-1-yl)benzaldehyde
Formula: C₁₂H₁₆N₂O
MW: 204.2682 g/mol
Melting Pt: 41…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00452233
CAS Number: 27913-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR