3-Amino-3,4-dihydro-4-oxo-N-[(tetrahydrofuran-2-yl)methyl]quinazoline-2-carboxamide

Supplier: Apollo Scientific
Warning

Synonyms: 3-Amino-3,4-dihydro-4-oxo-2-{[(tetrahydrofuran-2-yl)methyl]carbamoyl}quinazoline, 2-({[(3-Amino-3,4-dihydro-4-oxoquinazolin-2-yl)carbonyl]amino}methyl)furan

OR23565-250MG
APOSOR23565-250MGEA 63.8 EUR
APOSOR23565-250MG
3-Amino-3,4-dihydro-4-oxo-N-[(tetrahydrofuran-2-yl)methyl]quinazoline-2-carboxamide
3-Amino-3,4-dihydro-4-oxo-N-[(tetrahydrofuran-2-yl)methyl]quinazoline-2-carboxamide
Formula: C₁₄H₁₆N₄O₃
MW: 288,302 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD01566065
CAS Number: 646497-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR