3-Bromothiophenol 97%

Supplier: Apollo Scientific
Danger

Synonyms: m-Bromothiophenol, 3-Bromobenzenethiol, 1-Bromo-3-mercaptobenzene, 3-Bromobenzene-1-thiol

OR2454-1G OR2454-25G OR2454-5G
APOSOR2454-1GEA 23.2 EUR
APOSOR2454-1G APOSOR2454-25G APOSOR2454-5G
3-Bromothiophenol 97%
3-Bromothiophenol
Formula: C₆H₅BrS
Boiling Pt: 123…124 °C/40mmHg
Density: 1,832 g/cm³ (25 °C)
Flash Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009603
CAS Number: 6320-01-0
EINECS: 228-664-9
UN: 2922
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR