N-(tert-Butyl)hydroxylamine hydrochloride 95+%

Supplier: Apollo Scientific
Warning

OR24575-1G OR24575-5G OR24575-10G
APOSOR24575-1GEA 23.2 EUR
APOSOR24575-1G APOSOR24575-5G APOSOR24575-10G
N-(tert-Butyl)hydroxylamine hydrochloride 95+%
N-(tert-Butyl)hydroxylamine hydrochloride
Formula: C₄H₁₁NO
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00012598
CAS Number: 57497-39-9
EINECS: 260-771-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR