α-Chloro-m-xylene 94%

Supplier: Apollo Scientific
Danger

OR2562-100G OR2562-250G OR2562-1KG
APOSOR2562-100GEA 26.1 EUR
APOSOR2562-100G APOSOR2562-250G APOSOR2562-1KG
α-Chloro-m-xylene 94%
α-Chloro-m-xylene
Formula: C₈H₉Cl
Boiling Pt: 195…196 °C
Density: 1,064 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000909
CAS Number: 620-19-9
EINECS: 210-628-9
UN: 3265
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR