Tris(4-methylphenyl)phosphine 99%

Supplier: Apollo Scientific
Warning

OR25774-100G OR25774-25G OR25774-5G
APOSOR25774-100GEA 65.4 EUR
APOSOR25774-100G APOSOR25774-25G APOSOR25774-5G
Tris(4-methylphenyl)phosphine 99%
Tris(4-methylphenyl)phosphine
Formula: C₂₁H₂₁P
Melting Pt: 144…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00008542
CAS Number: 1038-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR