5-Chloro-3-pyridineboronic acid

Supplier: Apollo Scientific
Warning

OR2582-1G OR2582-25G OR2582-5G
APOSOR2582-1GEA 23.2 EUR
APOSOR2582-1G APOSOR2582-25G APOSOR2582-5G
5-Chloro-3-pyridineboronic acid
5-Chloro-3-pyridineboronic acid
Formula: C₅H₅BClNO₂
Melting Pt: 255…260 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06798243
CAS Number: 872041-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR