2-Carboxythiophene-5-boronic acid

Supplier: Apollo Scientific
Warning

OR25978-1G OR25978-250MG OR25978-5G
APOSOR25978-1GEA 27.7 EUR
APOSOR25978-1G APOSOR25978-250MG APOSOR25978-5G
2-Carboxythiophene-5-boronic acid
2-Carboxythiophene-5-boronic acid
Formula: C₅H₅BO₄S
Melting Pt: 135…136 °C
Storage Temperature: Ambient
MDL Number: MFCD02323211
CAS Number: 465515-31-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR