Ethyl-2-bromobenzoate

Supplier: Apollo Scientific
Warning

OR26844-25G OR26844-5G OR26844-100G
APOSOR26844-25GEA 24.8 EUR
APOSOR26844-25G APOSOR26844-5G APOSOR26844-100G
Ethyl-2-bromobenzoate
Ethyl 2-bromobenzoate
Formula: C₉H₉BrO₂
Boiling Pt: 255 °C
Density: 1,443 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00015443
CAS Number: 6091-64-1
EINECS: 228-034-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR