Benzamidoxime

Supplier: Apollo Scientific
Danger

OR27223-5G OR27223-1G OR27223-10G
APOSOR27223-5GEA 24.8 EUR
APOSOR27223-5G APOSOR27223-1G APOSOR27223-10G
Benzamidoxime
Benzamidoxime
Formula: C₇H₈N₂O
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00031485
CAS Number: 613-92-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR