4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one 98%

Supplier: Apollo Scientific

OR27385-25G OR27385-5G
APOSOR27385-25GEA 221 EUR
APOSOR27385-25G APOSOR27385-5G
4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one 98%
4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one
Formula: C₈H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00085024
CAS Number: 67751-23-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR