2,6-Dichloro-4-[5-(1,2-oxazol-5-yl)-1,3,4-oxadiazol-2-yl]pyridine

Supplier: Apollo Scientific
Warning

OR27433-250MG
APOSOR27433-250MGEA 79.9 EUR
APOSOR27433-250MG
2,6-Dichloro-4-[5-(1,2-oxazol-5-yl)-1,3,4-oxadiazol-2-yl]pyridine
2,6-Dichloro-4-[5-(1,2-oxazol-5-yl)-1,3,4-oxadiazol-2-yl]pyridine
Formula: C₁₀H₄Cl₂N₄O₂
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00106310
CAS Number: 680214-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR