6-Methyl-2-thiouracil

Supplier: Apollo Scientific
Warning

Synonyms: Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin

OR27585-50G
APOSOR27585-50GEA 21.9 EUR
APOSOR27585-50G
6-Methyl-2-thiouracil
6-Methyl-2-thiouracil
Formula: C₅H₆N₂OS
Melting Pt: 330 °C (dec.)
Density: 1,36 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006040
CAS Number: 56-04-2
EINECS: 200-252-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR