4-Chloro-2-cyclohexylphenol

Supplier: Apollo Scientific
Danger

OR28650-10G OR28650-1G OR28650-25G OR28650-100G OR28650-500G
APOSOR28650-10GEA 78.3 EUR
APOSOR28650-10G APOSOR28650-1G APOSOR28650-25G APOSOR28650-100G APOSOR28650-500G
4-Chloro-2-cyclohexylphenol
4-Chloro-2-cyclohexylphenol
Formula: C₁₂H₁₅ClO
MW: 210.7 g/mol
Boiling Pt: 170…180 °C/18mmHg
Melting Pt: 59…60 °C
Storage Temperature: Ambient
CAS Number: 13081-17-9
EINECS: 235-988-4
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR