3-Nitrophenyl isocyanate 97%

Supplier: Apollo Scientific
Danger

OR28931-25G OR28931-5G
APOSOR28931-25GEA 289 EUR
APOSOR28931-25G APOSOR28931-5G
3-Nitrophenyl isocyanate 97%
3-Nitrophenyl isocyanate

When melted, may contain small amount of sediment.

Formula: C₇H₄N₂O₃
Boiling Pt: 130…131 °C/11mmHg
Melting Pt: 51…52 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007220
CAS Number: 3320-87-4
EINECS: 222-025-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR